missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRPEL1 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97221AF488
This item is not returnable.
View return policy
Description
GRPEL1 Polyclonal antibody specifically detects GRPEL1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| GRPEL1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 488 | |
| FLJ25609, GREPEL1, grpE protein homolog 1, mitochondrial, GrpE-like 1, mitochondrial (E. coli), GrpE-like protein cochaperone, HMGEmt-GrpE#1, Mt-GrpE#1 | |
| Produced in rabbits immunized with E. coli-derived Human GRPEL1 fragment. | |
| 0.1 mL | |
| Endocrinology, Signal Transduction | |
| 80273 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction