missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GRPEL1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5109789

Product Code. 17117723

  • € 493.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MAAQCVRLAR RSLPALALSL RPSPRLLCTA TKQKNSGQNL EEDMGQSEQK ADPPATEKTL LEEKVKLEEQ LKETVEKYKR ALADTENLRQ RSQKLVEEAK LYGIQAFCKD LLEVADVLEK ATQCVPKEEI KDDNPHLKNL YEGLVMTEVQ IQKVFTKHGL LKLNPVGAKF DPYEHEALFH TPVEGKEPGT VALVSKVGYK LHGRTLRPAL VGVVKEA.

GRPEL1 (GrpE Like 1, Mitochondrial) is a Protein Coding gene. Diseases associated with GRPEL1 include Human Monocytic Ehrlichiosis and Lymphangitis. Among its related pathways are Metabolism of proteins and Mitochondrial protein import. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and chaperone binding. An important paralog of this gene is GRPEL2.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GRPEL1
Polyclonal
Unconjugated
GRPEL1
AA408748; FLJ25609; GREPEL1; GrpE; GrpE like 1, mitochondrial; grpE protein homolog 1, mitochondrial; GrpE#1; GrpE1; GRPEL1; GrpE-like 1; GrpE-like 1, mitochondrial; GrpE-like protein cochaperone; HMGE; mt-GrpE#1; mt-Grpel1; stress-inducible chaperone mt-GrpE#1
Rabbit
Affinity chromatography
RUO
17713, 79563, 80273
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P97576, Q99LP6, Q9HAV7
GRPEL1
Recombinant fusion protein containing a sequence corresponding to amino acids 1-217 of human GRPEL1 (NP_079472.1).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato