missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRPEL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557751
This item is not returnable.
View return policy
Description
Immunogen sequence: VLEKATQCVP KEEIKDDNPH LKNLYEGLVM TEVQIQKVFT KHGLLKLNPV GAKFDPYEHE ALFHTPVEGK EPGTVALVSK VGYK Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.
GRPEL1 (GrpE Like 1, Mitochondrial) is a Protein Coding gene. Diseases associated with GRPEL1 include Human Monocytic Ehrlichiosis and Lymphangitis. Among its related pathways are Metabolism of proteins and Mitochondrial protein import. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and chaperone binding. An important paralog of this gene is GRPEL2.
Specifications
| GRPEL1 | |
| Polyclonal | |
| Unconjugated | |
| GRPEL1 | |
| AA408748; FLJ25609; GREPEL1; GrpE; GrpE like 1, mitochondrial; grpE protein homolog 1, mitochondrial; GrpE#1; GrpE1; GRPEL1; GrpE-like 1; GrpE-like 1, mitochondrial; GrpE-like protein cochaperone; HMGE; mt-GrpE#1; mt-Grpel1; stress-inducible chaperone mt-GrpE#1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17713, 79563, 80273 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P97576, Q99LP6, Q9HAV7 | |
| GRPEL1 | |
| Recombinant protein corresponding to Human GRPEL1. Recombinant protein control fragment (Product #RP-96622). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction