missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GRRP1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15771021
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15771021 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15771021 Supplier Invitrogen™ Supplier No. PA553077

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: HQVIARRQEP ALRGSPGPLT PHPCNELGPP ASPRTPRPVR RGSGRRLPRP DSLIFYRQKR DC Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen GRRP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM110D
Gene Accession No. Q8TAY7
Gene Alias FAM110D; family with sequence similarity 110 member D; family with sequence similarity 110, member D; glycine/arginine rich protein 1; glycine/arginine-rich protein 1; GRRP1; Protein FAM110D
Gene Symbols FAM110D
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GRRP1. Recombinant protein control fragment (Product #RP-90785).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79927
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.