missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRSF1 Recombinant Rabbit Monoclonal Antibody (23GB1890), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 217.00 - € 590.00
Specifications
| Antigen | GRSF1 |
|---|---|
| Clone | 23GB1890 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087628
![]() |
Invitrogen™
MA554024 |
100 μL |
€ 590.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088639
|
Invitrogen™
MA554025 |
20 μL |
€ 217.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GRSF1 Recombinant Monoclonal antibody specifically detects GRSF1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| GRSF1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q12849 | |
| 2926 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1890 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| BB232551; C80280; D5Wsu31e; G-rich RNA sequence binding factor 1; G-rich sequence factor 1; Grsf1; GRSF-1 | |
| A synthesized peptide derived from human GRSF1 (350-400AA). | |
| Primary | |
| -20°C | |
| GRSF1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title