missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRSF1 Recombinant Rabbit Monoclonal Antibody (JE63-10)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544794
This item is not returnable.
View return policy
Description
GRSF1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
G-rich sequence factor 1 (GRSF1) is a regulator of post-transcriptional mitochondrial gene expression. It required for assembly of the mitochondrial ribosome and for recruitment of mRNS and IncRNA. GRSF1 binds RNAs containing the 14 base G-rich element. It preferentially binds RNAs transcribed from three contiguous genes on the light strand of mtDNA, the ND6 mRNA, and the long non-coding RNAs for MT-CYB and MT-NDS - each of which contains multiple consensus binding sequences.
Specifications
| GRSF1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q12849, Q8C5Q4 | |
| GRSF1 | |
| Synthetic peptide within human GRSF1 aa 101-150/480. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| JE63-10 | |
| Unconjugated | |
| GRSF1 | |
| BB232551; C80280; D5Wsu31e; G-rich RNA sequence binding factor 1; G-rich sequence factor 1; Grsf1; GRSF-1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 231413, 2926, 305256 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction