missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GRTP1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15739001 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15739001 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15739001 Supplier Invitrogen™ Supplier No. PA561182

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VALTLIKQHQ ELILEATSVP DICDKFKQIT KGSFVMECHT FMQKIFSEPG SLSMATVAKL RESCR Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 89%.

GRTP1, also known as germ cell-specific gene 1-related protein, is a protein that is expressed in the testis and is involved in the regulation of spermatogenesis, the process of sperm cell development. It is a member of the GRP family of proteins, which are characterized by a conserved motif called the GRP domain. GRTP1 is thought to play a role in the differentiation of spermatogonia into spermatocytes and in the regulation of meiosis, the process of cell division that produces haploid cells. Mutations in the GRTP1 gene have been associated with male infertility. GRTP1 was identified as an HIV dependency factor (HDF), suggesting that GRTP1 may be an important drug target in HIV treatment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRTP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GRTP1
Gene Accession No. Q5TC63
Gene Alias 5430401C05Rik; C81211; GH regulated TBC protein 1; growth hormone regulated TBC protein 1; growth hormone-regulated TBC protein 1; Grtp1; LRRGT00052; RP11-391H12.7; TBC1 domain family member 6; Tbc1d6
Gene Symbols GRTP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GRTP1. Recombinant protein control fragment (Product #RP-97976).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79774
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.