missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRTP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101613
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GRTP1.
GRTP1, also known as germ cell-specific gene 1-related protein, is a protein that is expressed in the testis and is involved in the regulation of spermatogenesis, the process of sperm cell development. It is a member of the GRP family of proteins, which are characterized by a conserved motif called the GRP domain. GRTP1 is thought to play a role in the differentiation of spermatogonia into spermatocytes and in the regulation of meiosis, the process of cell division that produces haploid cells. Mutations in the GRTP1 gene have been associated with male infertility. GRTP1 was identified as an HIV dependency factor (HDF), suggesting that GRTP1 may be an important drug target in HIV treatment.
Specifications
| GRTP1 | |
| Polyclonal | |
| Unconjugated | |
| GRTP1 | |
| 5430401C05Rik; C81211; GH regulated TBC protein 1; growth hormone regulated TBC protein 1; growth hormone-regulated TBC protein 1; Grtp1; LRRGT00052; RP11-391H12.7; TBC1 domain family member 6; Tbc1d6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 79774 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5TC63 | |
| GRTP1 | |
| A synthesized peptide derived from human GRTP1(Accession Q5TC63), corresponding to amino acid residues M79-T129. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction