missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540495
This item is not returnable.
View return policy
Description
Peptide sequence: FPALHGDSLY GASGGASSDY GAFYPRPVAP GGAGLPAAVS GSRLGYNNYF Sequence homology: Cow: 86%; Dog: 92%; Human: 100%; Mouse: 85%; Pig: 100%; Rat: 85%.
This gene encodes a member of the bicoid subfamily of the paired (PRD) homeobox family of proteins. The encoded protein acts as a transcription factor and may be autoregulatory. A similar protein in mice plays a role in craniofacial and rib cage development during embryogenesis.
Specifications
| GSC | |
| Polyclonal | |
| Unconjugated | |
| GSC | |
| GCS; goosecoid; goosecoid homeobox; goosecoid protein; GSC; Homeobox protein goosecoid; SAMS | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 145258 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P56915 | |
| GSC | |
| Synthetic peptide directed towards the N-terminal of human GSC (aa 35-84). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction