missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSDMB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562601
This item is not returnable.
View return policy
Description
Immunogen sequence: GKSLGSEDSR NMKEKLEDME SVLKDLTEEK RKDVLNSLAK CLGKEDIRQD LEQRVSEVLI SGELHMEDPD KPLLSSLFNA AGVLV Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 28%.
This gene encodes a member of the gasdermin-domain containing protein family. Other gasdermin-family genes are implicated in the regulation of apoptosis in epithelial cells, and are linked to cancer. Multiple transcript variants encoding different isoforms have been found for this gene. Additional variants have been described, but they are candidates for nonsense-mediated mRNA decay (NMD) and are unlikely to be protein-coding.
Specifications
| GSDMB | |
| Polyclonal | |
| Unconjugated | |
| GSDMB | |
| gasdermin B; gasdermin-B; Gasdermin-like protein; GSDMB; GSDML; PP4052; PRO2521 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55876 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TAX9 | |
| GSDMB | |
| Recombinant protein corresponding to Human GSDMB. Recombinant protein control fragment (Product #RP-93177). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction