missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSDMDC1 Antibody - Cleaved, N-terminal, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33474-100ul
This item is not returnable.
View return policy
Description
GSDMDC1 Monoclonal antibody specifically detects GSDMDC1 in Human samples. It is validated for ELISA,Western Blot
Specifications
| GSDMDC1 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| DF5L, DFNA5L, FLJ12150, gasdermin D, gasdermin domain containing 1, Gasdermin domain-containing protein 1, gasdermin-D, GSDMD N, GSDMDC1, GSDMD-N | |
| A synthetic peptide corresponding to residues surrounding Asp275 of human GSDMDC (NP_079012.3).,, Sequence:, NFLTD | |
| 100 μL | |
| Cell Biology | |
| 79792 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction