missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ GSDMDC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30246
This item is not returnable.
View return policy
Description
GSDMDC1 Polyclonal antibody specifically detects GSDMDC1 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| GSDMDC1 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| DF5L, DFNA5L, FLJ12150, gasdermin D, gasdermin domain containing 1, Gasdermin domain-containing protein 1, gasdermin-D, GSDMD N, GSDMDC1, GSDMD-N | |
| Between 434 and 484 | |
| 100 μg | |
| Cell Biology | |
| 79792 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction