missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114024
This item is not returnable.
View return policy
Description
Immunogen sequence: VLEFKCKHSK SFKENPNCLP HHHQCFPRRL SSAAPTVGPL TSYHQYHNQP For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.
May cause the redistribution of PAPOLB from the cytosol to the endoplasmic reticulum.
Specifications
| GSG1 | |
| Polyclonal | |
| Unconjugated | |
| GSG1 | |
| germ cell associated 1; germ cell-specific gene 1 protein; GSG1; testicular secretory protein Li 20; UNQ709/PRO1360 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 83445 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q2KHT4 | |
| GSG1 | |
| Synthetic peptide directed towards the C terminal region of human GSG1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction