missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSG1L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553177
This item is not returnable.
View return policy
Description
Immunogen sequence: QGYREEPTFI DPEAIKYFRE RMEKRDGSEE DFHLDCRHER YPARHQPHMA DSWPRSSAQE APELNRQCWV LGHWV Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 27%.
As a component of the inner core of AMPAR complex, modifies AMPA receptor (AMPAR) gating.
Specifications
| GSG1L | |
| Polyclonal | |
| Unconjugated | |
| GSG1L | |
| AI852434; C230098I05Rik; g1l; G630023A01Rik; germ cell-specific gene 1-like protein; Gm637; GSG1 like; GSG1L; GSG1-like; GSG1-like protein; KTSR5831; PRO19651; RGD1562278; UNQ5831/PRO19651 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 146395 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6UXU4 | |
| GSG1L | |
| Recombinant protein corresponding to Human GSG1L. Recombinant protein control fragment (Product #RP-91037). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction