missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK-3 alpha Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97994C
This item is not returnable.
View return policy
Description
GSK-3 alpha Polyclonal specifically detects GSK-3 alpha in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| GSK-3 alpha | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| DKFZp686D0638, EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 alpha, glycogen synthase kinase-3 alpha, GSK-3 alpha | |
| Produced in rabbits immunized with E. coli-derived Human GSK-3 alpha fragment. | |
| 0.1 mL | |
| Cell Biology, Hypoxia, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers | |
| 2931 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction