missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK-3 alpha/beta Antibody (1H8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-85523
This item is not returnable.
View return policy
Description
GSK-3 alpha/beta Monoclonal specifically detects GSK-3 alpha/beta in Human, Mouse, Rat, Porcine, Bovine, Canine, Hamster, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| GSK-3 alpha/beta | |
| Monoclonal | |
| Unconjugated | |
| DKFZp686D0638, EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 alpha, glycogen synthase kinase-3 alpha, GSK-3 alpha | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| GSK3 alpha/beta Antibody (1H8) | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1H8 | |
| Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| GSK3A | |
| Recombinant Xenopus GSK3 alpha/beta (1H8) | |
| 0.05 mg | |
| Cell Biology, Hypoxia, Protein Kinase, Signal Transduction, Stem Cell Markers | |
| 2931 | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Primate, Rabbit, Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction