missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK-3 alpha/beta, p Tyr216 Antibody (M132), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23367
This item is not returnable.
View return policy
Description
GSK-3 alpha/beta Monoclonal antibody specifically detects GSK-3 alpha/beta in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| GSK-3 alpha/beta | |
| Monoclonal | |
| Unconjugated | |
| PBS, 1 mg/ml BSA and 50% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| M132 | |
| Western Blot 1:1000 | |
| DKFZp686D0638, EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 alpha, glycogen synthase kinase-3 alpha, GSK-3 alpha | |
| Clone M132 was generated from a phospho-GSK-3 beta (Tyr-216) synthetic peptide (coupled to KLH) corresponding to amino acid residues around tyrosine 216 of human GSK-3 beta. This peptide sequence is also found in GSK-3 alpha (Tyr-279) and is highly conserved in GSK-3 genes in rat and mouse. [UniProt# P49840] | |
| 0.1 mL | |
| Cell Biology, Hypoxia, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers | |
| 2931 | |
| Store at -20°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction