missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK-3 beta Antibody (272536), DyLight 405, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB2506E
This item is not returnable.
View return policy
Description
GSK-3 beta Monoclonal antibody specifically detects GSK-3 beta in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Specifications
| GSK-3 beta | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 272536 | |
| Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK-3 beta, GSK3beta isoform | |
| E. coli-derived recombinant human GSK-3 beta , Met1-Thr420, Accession # P49841 | |
| 0.1 mL | |
| Cancer, Hypoxia, Lipid and Metabolism, mTOR Pathway, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
| 2932 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction