missing translation for 'onlineSavingsMsg'
Learn More

GSK 5072 Dihydrochloride, TRC

Product Code. 30003556
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003556 10 mg 10mg
30132745 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30003556 Supplier Toronto Research Chemicals Supplier No. TRCG79741510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C19H16ClN3S • 2(HCl)
Molecular Weight (g/mol) 353.8723646
Synonym 5-(((4-Chlorobenzyl)(pyridin-3-ylmethyl)amino)methyl)thiophene-2-carbonitrile Dihydrochloride, 5-[[[(4-Chlorophenyl)methyl](3-pyridinylmethyl)amino]methyl]-2-thiophenecarbonitrile Dihydrochloride;
SMILES ClC1=CC=C(C=C1)CN(CC2=CN=CC=C2)CC3=CC=C(C#N)S3.Cl.Cl

Specifications

Quantity 10 mg
Chemical Name or Material GSK 5072 Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.