Learn More
Tocris Bioscience™ GSK 6853
missing translation for 'orderingAttributeHoverText'
Quantity:
5 mg
25 mg
missing translation for 'unitSize'
25mg
5mg
Description
GSK 6853 is a potent and selective BRPF1 inhibitor. Exhibits >1600-fold selectivity for BRPF1 over other bromodomains (pIC50 values by TR-FRET are 8.1, 5.1/4.8 and 4.7/<4.3 for BRPF1, BRPF2/3 and BRD4 BD1/2, respectively).
Negative Control also available.
Specifications
Specifications
| Chemical Name or Material | N-[2,3-dihydro-1,3-dimethyl-6-[(2R)-2-methyl-1-piperazinyl]-2-oxo-1H-benzimidazol-5-yl]-2-methoxybenzamide |
| CAS | 1910124-24-1 |
| Quantity | 25 mg |
| Molecular Formula | C22H27N5O3 |
| Formula Weight | 409.48 |
| Purity | 98% |