missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK2018682, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
GSK2018682 is an agonist for S1P1 and S1P5 receptor with pEC50s of 7.7 and 7.2, respectively, and has no agonist activity towards human S1P2, S1P3, or S1P4. GSK2018682 is used in the research of multiple sclerosis.
Specifications
Specifications
| Chemical Name or Material | GSK2018682 |
| CAS | 1034688-30-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H21ClN4O4 |
| Quantity | 25 mg |
| Solubility Information | DMSO : ≥ 125 mg/mL (283.52 mM) |
| SMILES | O=C(O)CCCN1C=CC2=C1C=CC=C2C3=NOC(C4=CC(Cl)=C(OC(C)C)N=C4)=N3 |
| Molecular Weight (g/mol) | 440.88 |
| Formula Weight | 440.88 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction