Learn More
GSK2018682, MedChemExpress
Product Code. 16457626
Encompass
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16457626
Brand: Medchem Express HY19511
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress GSK2018682 is an agonist for S1P1 and S1P5 receptor with pEC50s of 7.7 and 7.2, respectively, and has no agonist activity towards human S1P2, S1P3, or S1P4. GSK2018682 is used in the research of multiple sclerosis.
GSK2018682 is an agonist for S1P1 and S1P5 receptor with pEC50s of 7.7 and 7.2, respectively, and has no agonist activity towards human S1P2, S1P3, or S1P4. GSK2018682 is used in the research of multiple sclerosis.
Specifications
Chemical Name or Material GSK2018682
CAS 1034688-30-6
Color White
Purity Grade Notes Research
Molecular Formula C22H21ClN4O4
Quantity 100 mg
Solubility Information DMSO : ≥ 125 mg/mL (283.52 mM)
SMILES O=C(O)CCCN1C=CC2=C1C=CC=C2C3=NOC(C4=CC(Cl)=C(OC(C)C)N=C4)=N3
Molecular Weight (g/mol) 440.88
Formula Weight 440.88
Percent Purity 98.25%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only