Learn More
GSK2193874, MedChemExpress
Product Code. 16459686
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
Product Code. 16459686
Brand: Medchem Express HY100720
MedChemExpress GSK2193874 is an orally active, potent, and selective TRPV4 antagonist with IC50s of 2 nM and 40 nM for rTRPV4 and hTRPV4.
GSK2193874 is an orally active, potent, and selective TRPV4 antagonist with IC50s of 2 nM and 40 nM for rTRPV4 and hTRPV4.
Specifications
Chemical Name or Material GSK2193874
CAS 1336960-13-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C37H38BrF3N4O
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (144.59 mM; Need ultrasonic)
SMILES O=C(C1=C(CN2CCC(N3CCCCC3)CC2)C(C4=CC=CC(C(F)(F)F)=C4)=NC5=CC(Br)=CC=C15)NC6(C7=CC=CC=C7)CC6
Molecular Weight (g/mol) 691.62
Formula Weight 691.62
Percent Purity 98.33%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only