missing translation for 'onlineSavingsMsg'
Learn More

GSK2983559 active metabolite, MedChemExpress

Product Code. 16447898
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16447898

Brand: Medchem Express HY19764

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK2983559 active metabolite is an active metabolite of GSK2983559. GSK2983559 active metabolite is a receptor interacting protein-2 (RIP2) kinase inhibitor extracted from patent WO/2014043446 A1, compound example 1.

GSK2983559 active metabolite is an active metabolite of GSK2983559. GSK2983559 active metabolite is a receptor interacting protein-2 (RIP2) kinase inhibitor extracted from patent WO/2014043446 A1, compound example 1.

Specifications

Chemical Name or Material GSK2983559 active metabolite
CAS 1423186-80-4
Color Gray
Purity Grade Notes Research
Molecular Formula C21H22N4O4S2
Quantity 10 mg
Solubility Information DMSO : 25 mg/mL (54.52 mM; Need ultrasonic)
SMILES CC(C)(C)S(C1=C(OCCO)C=C(N=CN=C2NC3=CC(N=CS4)=C4C=C3)C2=C1)(=O)=O
Molecular Weight (g/mol) 458.55
Formula Weight 458.55
Percent Purity 98.24%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.