missing translation for 'onlineSavingsMsg'
Learn More

GSK3179106, MedChemExpress

Product Code. 16414336
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16414336

Brand: Medchem Express HY100459

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK3179106 is an orally active and selective RET kinase inhibitor with IC50s of 0.4 nM, 0.2 nM for human RET and rat RET, respectively. GSK3179106 has the potential for irritable bowel syndrome (IBS) through the attenuation of post-inflammatory and stress-induced visceral hypersensitivity.

GSK3179106 is an orally active and selective RET kinase inhibitor with IC50s of 0.4 nM, 0.2 nM for human RET and rat RET, respectively. GSK3179106 has the potential for irritable bowel syndrome (IBS) through the attenuation of post-inflammatory and stress-induced visceral hypersensitivity.

Specifications

Chemical Name or Material GSK3179106
CAS 1627856-64-7
Color White
Purity Grade Notes Research
Molecular Formula C22H21F4N3O4
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (213.94 mM)
SMILES O=C(NC1=NOC(C(C)(C)C(F)(F)F)=C1)CC2=CC=C(C(C(OCC)=C3)=CNC3=O)C=C2F
Molecular Weight (g/mol) 467.41
Formula Weight 467.41
Percent Purity 99.4%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.