missing translation for 'onlineSavingsMsg'
Learn More

GSK3787, MedChemExpress

Product Code. 16496638
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496638 10 mg 10mg
16412737 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16496638 Supplier Medchem Express Supplier No. HY15577

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK3787 is a selective and irreversible peroxisome proliferator-activated receptor δ (PPARδ) antagonist with pIC50 of 6.6.

GSK3787 is a selective and irreversible peroxisome proliferator-activated receptor δ (PPARδ) antagonist with pIC50 of 6.6.

Specifications

Chemical Name or Material GSK3787
Physical Form Solid
CAS 188591-46-0
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C15H12ClF3N2O3S
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 50 mg/mL (127.30 mM)
SMILES O=C(NCCS(=O)(C1=NC=C(C(F)(F)F)C=C1)=O)C2=CC=C(Cl)C=C2
Molecular Weight (g/mol) 392.78
Formula Weight 392.78
Percent Purity 99.04%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.