Learn More
GSNOR,ADH5 Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-16379-100UL
Description
The GSNOR,ADH5 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human GSNOR,ADH5. This antibody recognizes human, mouse, rat antigen. The GSNOR,ADH5 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™488.
Specifications
| GSNOR,ADH5 | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat |
| Flow Cytometry (Intracellular Staining) | |
| CoraLite 488 | |
| 100 μL | |
| 128 | |
| IgG |