Learn More
GSNOR ADH5 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-11051-100UL
Description
The GSNOR,ADH5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GSNOR,ADH5. This antibody recognizes human, mouse, rat antigen. The GSNOR,ADH5 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
| GSNOR ADH5 | |
| 1000 μg/mL | |
| ADH 3, ADH5, ADHX, Alcohol dehydrogenase 5, Alcohol dehydrogenase class 3, FALDH, FDH, GSH FDH, GSNOR | |
| 100 μL | |
| 128 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry (Intracellular Staining) | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |