Learn More
GSNOR ADH5 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-11051-100UL
 View more versions of this product
Product Code. 18028935
  • € 450.00 / 100µL
Description
The GSNOR,ADH5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GSNOR,ADH5. This antibody recognizes human, mouse, rat antigen. The GSNOR,ADH5 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
GSNOR ADH5
1000 μg/mL
ADH 3, ADH5, ADHX, Alcohol dehydrogenase 5, Alcohol dehydrogenase class 3, FALDH, FDH, GSH FDH, GSNOR
100 μL
128
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Flow Cytometry (Intracellular Staining)
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa