missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSPT2 Mouse anti-Human, Clone: OTI2G9, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503066S
This item is not returnable.
View return policy
Description
ERF3B encodes a GTPase that belongs to the GTP-binding elongation factor family. The encoded protein is a polypeptide release factor that complexes with eukaryotic peptide chain release factor 1 to mediate translation termination. This protein may also be involved in mRNA stability.Specifications
| GSPT2 | |
| Monoclonal | |
| 0.32 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8IYD1 | |
| GSPT2 | |
| Full length human recombit protein of human GSPT2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI2G9 | |
| Unconjugated | |
| GSPT2 | |
| ERF3B, GST2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23708 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction