missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSTA4 Polyclonal Antibody

Rabbit Polyclonal Antibody
€ 180.00 - € 476.00
Specifications
| Antigen | GSTA4 |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17770554
![]() |
Invitrogen™
PA5119145 |
200 μL |
€ 476.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17793334
|
Invitrogen™
PA5121941 |
20 μL |
€ 180.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GSTA4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Conjugation of reduced glutathione to a wide number of exogenous and endogenous hydrophobic electrophiles. This isozyme has a high catalytic efficiency with 4-hydroxyalkenals such as 4-hydroxynonenal (4-HNE).Specifications
| GSTA4 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Rat | |
| GSTA4 | |
| glutathione S-alkyltransferase A4; glutathione S-aralkyltransferase A4; glutathione S-aryltransferase A4; glutathione S-transferase 5.7; Glutathione S-transferase A4; glutathione S-transferase A4-4; glutathione S-transferase alpha 4; glutathione S-transferase, alpha 4; glutathione transferase A4-4; GST 5.7; GST A4-4; GST class-alpha member 4; GST5.7; Gsta; GSTA4; GSTA4-4; GTA4; mGsta4; S-(hydroxyalkyl)glutathione lyase A4 | |
| GSTA4 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P14942 | |
| 300850 | |
| Recombinant protein Glutathione S Transferase A4. The antigen corresponds to amino acid range 2-222 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title