missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTM2 Antibody (CPTC-GSTMu2-2), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08255AF750
This item is not returnable.
View return policy
Description
GSTM2 Monoclonal antibody specifically detects GSTM2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| GSTM2 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| glutathione S-transferase mu 2 (muscle), MGC117303, muscle, S-(hydroxyalkyl)glutathione lyase M2 | |
| Recombinant human full-length GSTM2 protein (Uniprot: P28161) | |
| 100 μg | |
| Endocrinology, Signal Transduction | |
| 2946 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CPTC-GSTMu2-2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction