missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTM2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 326.00
Specifications
| Antigen | GSTM2 |
|---|---|
| Dilution | Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18652387
|
Novus Biologicals
NBP2-99918-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18675997
|
Novus Biologicals
NBP2-99918-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GSTM2 Polyclonal antibody specifically detects GSTM2 in Human samples. It is validated for Western Blot, ELISASpecifications
| GSTM2 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Endocrinology, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 2946 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| glutathione S-transferase mu 2 (muscle), MGC117303, muscle, S-(hydroxyalkyl)glutathione lyase M2 | |
| Produced in rabbits immunized with purified, recombinant Human GSTM2 (Accession#: P28161; Met 1-Lys 218) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title