missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTM3 Antibody (20C11.H5.E11), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-44175UV
This item is not returnable.
View return policy
Description
GSTM3 Monoclonal antibody specifically detects GSTM3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| GSTM3 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 2947 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry | |
| 20C11.H5.E11 | |
| Western Blot, ELISA, Immunohistochemistry | |
| glutathione S-transferase mu 3 (brain), GST5Mu-3, MGC3310, MGC3704, S-(hydroxyalkyl)glutathione lyase M3 | |
| This Protein A purified GSTM3 Antibody (20C11.H5.E11) was prepared by repeated immunizations in mice with GSTGSTM3 Protein (Uniprot: P21266) | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction