missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTM3 Antibody (rGSTM3/8878), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24085JF549
This item is not returnable.
View return policy
Description
GSTM3 Monoclonal antibody specifically detects GSTM3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| GSTM3 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| glutathione S-transferase mu 3 (brain), GST5Mu-3, MGC3310, MGC3704, S-(hydroxyalkyl)glutathione lyase M3 | |
| Recombinant full-length human GSTM3 protein | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 2947 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| rGSTM3/8878 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction