missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSTM4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121939
This item is not returnable.
View return policy
Description
GSTM4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: alpha, kappa, mu, omega, pi, sigma, theta and zeta. This gene encodes a glutathione S-transferase that belongs to the mu class. The mu class of enzymes functions in the detoxification of electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins and products of oxidative stress, by conjugation with glutathione. The genes encoding the mu class of enzymes are organized in a gene cluster on chromosome 1p13.3 and are known to be highly polymorphic. These genetic variations can change an individual's susceptibility to carcinogens and toxins as well as affect the toxicity and efficacy of certain drugs. Diversification of these genes has occurred in regions encoding substrate-binding domains, as well as in tissue expression patterns, to accommodate an increasing number of foreign compounds. Multiple transcript variants, each encoding a distinct protein isoform, have been identified.
Specifications
| GSTM4 | |
| Polyclonal | |
| Unconjugated | |
| GSTM4 | |
| 1110004G14Rik; glutathione S-alkyltransferase M4; glutathione S-aralkyltransferase M4; glutathione S-aryltransferase M4; glutathione S-transferase M4; glutathione S-transferase mu 3; glutathione S-transferase Mu 4; glutathione S-transferase, mu 4; glutathione-S-transferase, mu 4; GST class-mu 4; Gstb4; Gstb-4; GSTM4; GSTM4-4; GSTM7-7; GST-Mu2; GTM4; GTS-Mu2; S-(hydroxyalkyl)glutathione lyase M4; testis tissue sperm-binding protein Li 60n | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 499689 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| 0 | |
| GSTM4 | |
| Recombinant protein Glutathione S Transferase Mu 4. | |
| 20 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction