missing translation for 'onlineSavingsMsg'
Learn More

GSTO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. p-7206730 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16834164 20 μL 20µL
16824164 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16834164 Supplier Proteintech Supplier No. 151241AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody detects both monomer (27-30 kDa) and dimer forms of GSTO1 (54-64 kDa).

GSTO1 (glutathione S-transferase omega 1), also known as p28 or GSTTLp28, is a 241 amino acid protein that localizes to the cytoplasm and contains both an N-terminal and a C-terminal GST domain. Expressed ubiquitously with highest expression in heart, liver and skeletal muscle, GSTO1 exists as a homodimer that functions as both a glutathione-dependent thiol transferase and a dehydroascorbate reductase. Specifically, GSTO1 catalyzes the reaction of glutathione with a wide variety of organic compounds to form thioethers, a process that is essential for the metabolism and detoxification of a variety of xenobiotics and carcinogens. Human GSTO1 shares 70% sequence homology with its rodent counterpart, suggesting a conserved role between species. Polymorphisms in the gene encoding GSTO1 may be associated with the development of childhood acute lymphoblastic leukemia, Parkinson′s disease and Alzheimer disease.

Specifications

Antigen GSTO1
Applications Immunoprecipitation, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GSTO1
Gene Accession No. O09131, P78417, Q9Z339
Gene Alias GSTO 1, GSTO1, GSTTLp28, P28
Gene Symbols GSTO1
Host Species Rabbit
Immunogen GSTO1 Fusion Protein Ag7199
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114846, 14873, 9446
Target Species Mouse, Rat, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.