missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSTP1 Monoclonal Antibody (1A7B5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA538533
This item is not returnable.
View return policy
Description
This antibody has been tested in indirect ELISA.
Glutathione S-transferases (GSTs) are a family of enzymes that play an important role in detoxification by catalyzing the conjugation of many hydrophobic and electrophilic compounds with reduced glutathione. Based on their biochemical, immunologic, and structural properties, the soluble GSTs are categorized into 4 main classes: alpha, mu, pi, and theta. This GST family member is a polymorphic gene encoding active, functionally different GSTP1 variant proteins that are thought to function in xenobiotic metabolism and play a role in susceptibility to cancer, and other diseases.
Specifications
| GSTP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P09211 | |
| Gstp1 | |
| Purified recombinant fragment of human GSTP1 (aa: 2-210) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1A7B5 | |
| Unconjugated | |
| Gstp1 | |
| Chain 7; deafness, X-linked 7; DFN7; epididymis secretory protein Li 22; FAEES3; fatty acid ethyl ester synthase III; glutathione S-transferase P; Glutathione S-transferase P 1; glutathione S-transferase pi 1; glutathione S-transferase pi 2; glutathione S-transferase, pi 1; glutathione S-transferase, pi 2; Glutathione-S-transferase placental enzyme pi type; glutathione-S-transferase, pi 1; Glutathione-S-transferase, placental enzyme pi type; GST 7-7; GST class-pi; gst P1; Gst p-1; GST YF-YF; Gst3; Gstp; GST-P; Gstp1; GSTP1-1; Gstp2; GstpiB; GST-piB; HEL-S-22; HGNC:4638; PI; Preadipocyte growth factor | |
| Mouse | |
| Protein G | |
| RUO | |
| 2950 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction