missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTT2 Antibody (OTI3B6), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70861PE
This item is not returnable.
View return policy
Description
GSTT2 Monoclonal specifically detects GSTT2 in Human samples. It is validated for Flow Cytometry.
Specifications
| GSTT2 | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| GSTT2 | |
| Full length recombinant protein of human GSTT2 (NP_000845) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| OTI3B6 | |
| Flow Cytometry | |
| EC 2.5.1.18, glutathione S-transferase theta 2, glutathione S-transferase theta-2, GST class-theta-2, MGC182032 | |
| Mouse | |
| Affinity Purified | |
| Cancer | |
| 2953.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction