Learn More
GSTT2 Monoclonal Antibody (OTI5A11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA501933S
Encompass_Preferred
 View more versions of this product
Product Code. 17515551
  • € 242.00 / 30µL
Description
GSTT2 Monoclonal antibody specifically detects GSTT2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
GSTT2
Monoclonal
0.99 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P48637
Mouse
Affinity chromatography
RUO
2937
-20° C, Avoid Freeze/Thaw Cycles
IgG2b
Immunohistochemistry (Paraffin), Western Blot
OTI5A11
Unconjugated
GSS
AI266894; Glutathione S-transferase 12; glutathione S-transferase theta 2 (gene/pseudogene); glutathione S-transferase theta-2; Glutathione S-transferase Yrs-Yrs; glutathione S-transferase, theta 2; GST 12-12; GST class-theta-2; GSTT2; mGSTT2; Yrs
Full length human recombinant protein of human GSS produced in HEK293T cell.