missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSTT2B Monoclonal Antibody (K24025_6G4), Invitrogen™
Mouse Monoclonal Antibody
€ 450.00 - € 676.00
Specifications
| Antigen | GSTT2B |
|---|---|
| Clone | K24025_6G4 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30200297
|
Invitrogen™
MA556028 |
200 μL |
€ 676.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30206754
|
Invitrogen™
MA555648 |
50 μL |
€ 450.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: SQPSRAVYIF AKKNGIPLEL RTVDLVKGQH KSKEFLQINS LGKLPTLKDG DFILTESSAI LIYLSCKYQT PDHWYPSDLQ ARARVHEYLG WHADCIRGTF GIPLWVQVLG PLIGVQVPEE KVERNRTAMD QALQWLEDKF LGDRPFLAGQ QVTLADLMAL EELMQPVALG YELFEGRPRL AAWRGRVEAF LGAELCQEAH SIILSILEQA AKKTLPTPSP
GSTT2B Monoclonal antibody specifically detects GSTT2B in Human, Rat samples. It is validated for Western BlotSpecifications
| GSTT2B | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| glutathione S-transferase theta 2; glutathione S-transferase theta 2B (gene/pseudogene); glutathione S-transferase theta-2; Glutathione S-transferase theta-2B; GST class-theta-2; GSTT2; GSTT2B; GSTT2P | |
| Recombinant human GSTT2B (immunogen range is 11-230). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| GSTT2B |
| K24025_6G4 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| P0CG30 | |
| 653689 | |
| IgG2b | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title