missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTT2B Polyclonal Antibody

GSTT2B Polyclonal Antibody
Brand: Invitrogen PA5109794
This item is not returnable.
View return policy
Description
GSTT2B Polyclonal Antibody for Western Blot
Specifications
| GSTT2B | |
| Polyclonal | |
| Unconjugated | |
| GSTT2B | |
| glutathione S-transferase theta 2; glutathione S-transferase theta 2B (gene/pseudogene); glutathione S-transferase theta-2; Glutathione S-transferase theta-2B; GST class-theta-2; GSTT2; GSTT2B; GSTT2P | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 653689 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.98 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P0CG30 | |
| GSTT2B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 115-244 of human GSTT2B (NP_001074312.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction