Learn More
GSTZ1 Antibody (24E9.G9.G8), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-44177APC
Product Code. 18645945
  • € 724.00 / 0.10mL
Description
GSTZ1 Monoclonal antibody specifically detects GSTZ1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, SDS-Page
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GSTZ1
Monoclonal
APC
EC 2.5.1.18, EC 5.2.1.2, glutathione S-alkyltransferase, glutathione S-aralkyltransferase, Glutathione S-transferase zeta 1, glutathione transferase zeta 1, GSTZ1-1maleylacetoacetate isomerase, MAAIglutathione S-aryltransferase, MAI, maleylacetone isomerase, MGC2029, S-(hydroxyalkyl)glutathione lyase
This Protein A purified GSTZ1 Antibody (24E9.G9.G8) was prepared by repeated immunizations in mice with GSTGSTZ1 Protein (Uniprot: O43708)
0.1 mL
metabolism
2954
Store at 4°C in the dark.
IgG1 κ
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, SDS-Page
24E9.G9.G8