Learn More
GTF2A1L Mouse anti-Human, Clone: OTI4B4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF810961
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The assembly and stability of the RNA polymerase II transcription pre-initiation complex on a eukaryotic core promoter involve the effects of TFIIA on the interaction between TATA-binding protein (TBP) and DNA. This gene encodes a germ cell-specific counterpart of the large (alpha/beta) subunit of general transcription factor TFIIA that is able to stabilize the binding of TBP to DNA and may be uniquely important to testis biology. Alternative splicing for this locus has been observed and two variants, encoding distinct isoforms, have been identified. Co-transcription of this gene and the neighboring upstream gene generates a rare transcript (SALF), which encodes a fusion protein comprised of sequence sharing identity with each individual gene product.Specifications
| GTF2A1L | |
| Monoclonal | |
| Unconjugated | |
| GTF2A1L | |
| ALF | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 11036 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI4B4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9UNN4 | |
| GTF2A1L | |
| Human recombit protein fragment corresponding to amino acids 183-478 of human GTF2A1L produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.