missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ GTF2H1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29827
This item is not returnable.
View return policy
Description
GTF2H1 Polyclonal antibody specifically detects GTF2H1 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| GTF2H1 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Basic transcription factor 2 62 kDa subunit, BTF2 p62, BTF2general transcription factor IIH, polypeptide 1 (62kD subunit), General transcription factor IIH polypeptide 1, general transcription factor IIH subunit 1, general transcription factor IIH, polypeptide 1, 62kDa, TFB1, TFIIH, TFIIH basal transcription factor complex p62 subunit | |
| between 250 and 300 | |
| 100 μg | |
| Cell Cycle and Replication, DNA Repair, Nucleotide Excision Repair | |
| 2965 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction