missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF2H3 Monoclonal Antibody (OTI6E12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812725
This item is not returnable.
View return policy
Description
GTF2H3 interacts with HIV-1 Tat as a component of the HIV-1 transcription pre-initiation complex, but is released from the elongation complex which includes P-TEFb. It synergizes with HIV-1 Tat to induce transcription elongation from the HIV-1 LTR promoter.Specifications
| GTF2H3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13889 | |
| GTF2H3 | |
| Human recombit protein fragment corresponding to amino acids 1-241 of human GTF2H3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI6E12 | |
| Unconjugated | |
| GTF2H3 | |
| BTF2, P34, TFB4, TFIIH | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2967 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction