missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF2H3 Monoclonal Antibody (OTI6E12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812725S
This item is not returnable.
View return policy
Description
GTF2H3 Monoclonal antibody specifically detects GTF2H3 in Human samples. It is validated for Western BlotSpecifications
| GTF2H3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13889 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2967 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI6E12 | |
| Unconjugated | |
| GTF2H3 | |
| 34kDa; 5033417D07Rik; Basic transcription factor 2 34 kDa subunit; BTF2; BTF2 p34; BTF2-p34; C730029A10; D5Ertd679e; General transcription factor IIH polypeptide 3; general transcription factor IIH subunit 3; general transcription factor IIH, polypeptide 3; general transcription factor IIH, polypeptide 3, 34kDa; Gtf2h3; P34; TFB4; TFIIH; TFIIH basal transcription factor complex p34 subunit | |
| Human recombinant protein fragment corresponding to amino acids 1-241 of human GTF2H3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction