Learn More
GTF2H3 Mouse anti-Human, Clone: OTI6E12, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF812725
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
GTF2H3 interacts with HIV-1 Tat as a component of the HIV-1 transcription pre-initiation complex, but is released from the elongation complex which includes P-TEFb. It synergizes with HIV-1 Tat to induce transcription elongation from the HIV-1 LTR promoter.Specifications
| GTF2H3 | |
| Monoclonal | |
| Unconjugated | |
| GTF2H3 | |
| BTF2, P34, TFB4, TFIIH | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2967 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI6E12 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q13889 | |
| GTF2H3 | |
| Human recombit protein fragment corresponding to amino acids 1-241 of human GTF2H3 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.