missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GTF2I Recombinant Rabbit Monoclonal Antibody (HL1674)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547136
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
This gene encodes a multifunctional phosphoprotein with roles in transcription and signal transduction. It is deleted in Williams-Beuren syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at chromosome 7q11. 23. Alternative splicing results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 7, 13 and 21.
Specifications
| GTF2I | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P78347 | |
| GTF2I | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human TFII-I. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL1674 | |
| Unconjugated | |
| GTF2I | |
| 6030441I21Rik; BAP135; BAP-135; Bruton tyrosine kinase-associated protein 135; BTKAP1; BTK-associated protein 135; BTK-associated protein, 135kD; DIWS; Diws1t; general transcription factor II I; general transcription factor IIi; general transcription factor II-I; Gtf2i; Gtf2ird1; GTFII-I; IB291; OTTHUMP00000209509; Spin; SRF-Phox1-interacting protein; tfiii; TFII-I; WBS; WBSCR6; Williams-Beuren syndrome chromosomal region 6 protein; Williams-Beuren syndrome chromosome region 6 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2969 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction