missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF2I, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00002969-D01.100uL
This item is not returnable.
View return policy
Description
Sequence: MAQVAMSTLPVEDEESSESRMVVTFLMSALESMCKELAKSKAEVACIAVYETDVFVVGTERGRAFVNTRKDFQKDFVKYCVEEEEKAAEMHKMKSTTQANRMSVDAVEIETLRKTVEDYFCFCYGKALGKSTVVPVPYEKMLRDQSAVVVQGLPEGVAFKHPENYDLATLKWILENKAGISFIIKRPFLEPKKHVGGRVMVTDADRSILSPGGSCGPIKVKTEPTEDSGISLEMAAVTVKEESEDPDYYQYNIQGPSETDDVDEKQPLSKPLQGSHHSSEGNEGTEMEVPAEDDDYSPPSKRPKANELPQPPVPEPANAGKRKVREFNFEKWNARITDLRKQVEELFERKYAQAIKAKGPVTIPYPLFQSHVEDLYVEGLPEGIPFRRPSTYGIPRLERILLAKERIRFVIKKHELLNSTREDLQLDKPASGVKEEWYARITKLRKMVDQLFCKKFAEALGSTEAKAVPYQKFEAHPNDLYVEGLPENIPFRSPSWYGIPRLEKIIQVGNRIKFVIKRPELLTHSTTEVTQPRTNTPVKEDWNVRITKLRKQVEEIFNLKFAQALGLTEAVKVPYPVFESNPEFLYVEGLPEGIPFRSPTWFGIPRLERIVRGSNKIKFVVKKPELVISYLPPGMASKINTKALQSPKRPRSPGSNSKVPEIEVTVEGPNNNNPQTSAVRTPTQTNGSNVPFKPRGREFSFEAWNAKITDLKQKVENLFNEKCGEALGLKQAVKVPFALFESFPEDFYVEGLPEGVPFRRPSTFGIPRLEKILRNKAKIKFIIKKPEMFETAIKESTSSKSPPRKINSSPNVNTTASGVEDLNIIQVTIPDDDNERLSKVEKARQLREQVNDLFSRKFGEAIGMGFPVKVPYRKITINRGCVVVDGMPPGVSFKAPSYLEISSMRRILDSAEFIKFTVIRPFPGLVVNNQLVDQSESEGPVIQESAEPSQLEVPATEEIKETDGSSQIKQEPDPTWSpecifications
| GTF2I | |
| Polyclonal | |
| No additive | |
| BC099907.1 | |
| GTF2I | |
| GTF2I (AAH99907.1, 1 a.a. ∼ 976 a.a) full-length human protein. | |
| RUO | |
| 2969 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunoprecipitation | |
| Unconjugated | |
| general transcription factor II, i | |
| BAP-135/BAP135/BTKAP1/DIWS/FLJ38776/FLJ56355/IB291/SPIN/TFII-I/WBS/WBSCR6 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction