missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF2IRD2 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00084163-B01P
This item is not returnable.
View return policy
Description
GTF2IRD2 Polyclonal antibody specifically detects GTF2IRD2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
| GTF2IRD2 | |
| Polyclonal | |
| PBS (pH 7.4) | |
| FLJ21423, FP630, general transcription factor II i repeat domain 2 alpha, general transcription factor II-I repeat domain-containing protein 2A, GTF2I repeat domain containing 2, GTF2I repeat domain-containing protein 2A, GTF2IRD2 alpha, GTF2IRD2AFLJ37938, MGC75203, transcription factor GTF2IRD2, Transcription factor GTF2IRD2-alpha | |
| GTF2IRD2 (AAH67859, 1 a.a. - 498 a.a.) full-length human protein. MAQVAVSTLPVEEESSSETRMVVTFLVSALESMCKELAKSKAEVACIAVYETDVFVVGTERGCAFVNARTDFQKDFAKYCVAEGLCEVKPPCPVNGMQVHSGETEILRKAVEDYFCFCYGKALGTTVMVPVPYEKMLRDQSAVVVQGLPEGVAFQHPENYDLATLKWILENKAGISFIINRPFLGPESQLGGPGMVTDAERSIVSPSESCGPINVKTEPMEDSGISLKAEAVSVKKESEDPNYYQYNMQGSHPSSTSNEVIEMELPMEDSTPLVPSEEPNEDPEAEVKIEGNTNSSSVTNSAAGVEDLNIVQVTVPDNEKERLSSIEKIKQLREQVNDLFSRKFGEAIGVDFPVKVPYRKITFNPGCVVIDGMPPGVVFKAPGYLEISSMRRILEAAEFIKFTVIRPLPGLELSNGLLEAGAGGLLEAGSLRPAWATWQDPISTKNLKISWKQWLTPVAPATWQAEAGGSLELRSLRQQRTIIAILHSSLGDRTRPCL | |
| 50 μg | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| AAH67859 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 84163 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction